Learn More
Meteorin Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3475
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418194
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Meteorin Polyclonal antibody AF3475 Specifically detects Meteorin in Mouse samples. It is validated for Immunocytochemistry, Western Blotting
Spezifikation
Spezifikation
Meteorin
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q8C1Q4
Metrn
Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1, Gly22-Asp291, Accession # Q8C1Q4
Immunogen affinity purified
RUO
Primary
79006
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Meteorin in direct ELISAs and Western blots.