Learn More
Prostasin/Prss8 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2968
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417904
  • 360.37 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Prostasin/Prss8 Polyclonal antibody AF2968 Specifically detects Prostasin/Prss8 in Mouse samples. It is validated for ELISA Capture, Western Blotting
Spezifikation
Spezifikation
Prostasin/Prss8
LYOPH
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Q99L44
CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, prostasin, protease, serine, 8, Prss8, Serine protease 8
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Prostasin/Prss8 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity is observed with recombinant human Prostasin.
Mouse
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PRSS8
Mouse myeloma cell line NS0-derived recombinant mouse Prostasin/Prss8, Ala30-Gln289, Accession # Q99L44
Immunogen affinity purified
RUO
Primary
5652
Reconstitute at 0.2 mg/mL in sterile PBS.