Learn More
Splunc2 Monoclonal Antibody (1F12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517126
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17126 targets Splunc2 in FACS, ICC, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17126 immunogen is purified recombinant fragment of mouse Splunc2 expressed in E. Coli.
Splunc2 Monoclonal antibody specifically detects Splunc2 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Splunc2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P07743, Q96DR5 | |
BPIFA2 | |
Purified recombinant fragment of mouse Splunc2 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1F12 | |
Unconjugated | |
BPIFA2 | |
bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; BPIFA2; Bpifa2e; C20orf70; mSplunc2; parotid secretory protein; PSP; short palate; short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025 | |
Mouse | |
Protein G | |
RUO | |
140683, 19194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |