Learn More
TIM-3 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1529
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416994
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
TIM-3 Polyclonal antibody AF1529 Specifically detects TIM-3 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Spezifikation
Spezifikation
TIM-3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
AAL65156
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse TIM-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human TIM-3 is observed and less than 5% cross-reactivity with recombinant mouse (rm) TIM-1, rmTIM-2, rmTIM-4, rmTIM-5, rmTIM-6, and rmTIM-7 is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HAVCR2
Mouse myeloma cell line NS0-derived recombinant mouse TIM-3, Leu22-Arg191, Accession # AAL65156
Immunogen affinity purified
RUO
Primary
84868
Reconstitute at 0.2 mg/mL in sterile PBS.