Learn More
TIM-6 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2737
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417754
  • 324.73 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
TIM-6 Polyclonal antibody AF2737 Specifically detects TIM-6 in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
TIM-6
LYOPH
Western Blot 0.1 ug/mL
NP_001020606
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse TIM-6, Glu24-Ala140, Accession # NP_001020606
Immunogen affinity purified
RUO
Primary
Detects mouse TIM-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human TIM-3 and recombinant mouse TIM-4 is observed.