Learn More
TREM-3 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2224
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417424
  • 324.73 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
TREM-3 Polyclonal antibody AF2224 Specifically detects TREM-3 in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
TREM-3
LYOPH
Western Blot 0.1 ug/mL
NP_067382
Trem3
Mouse myeloma cell line NS0-derived recombinant mouse TREM-3 Gly20-Pro135 Accession # NP_067382
Immunogen affinity purified
RUO
Primary
54210
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse TREM-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) TREM-2b and rmTREM-1 is observed.