Learn More
c-Mpl Monoclonal Antibody (1H2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517127
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17127 targets MPL in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-17127 immunogen is purified recombinant fragment of human MPL expressed in E. Coli. MA5-17127 detects MPL which has a predicted molecular weight of approximately 71.2kDa.
c-Mpl Monoclonal antibody specifically detects c-Mpl in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.Spezifikation
c-Mpl | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P40238 | |
MPL | |
Purified recombinant fragment of human MPL expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Western Blot | |
1H2 | |
Unconjugated | |
MPL | |
CD110; c-mpl; hlb219; MPL; MPL proto-oncogene, thrombopoietin receptor; MPLV; myeloproliferative leukemia protein; myeloproliferative leukemia virus oncogene; proto-oncogene c-Mpl; THCYT2; thrombopoietin receptor; TPOR; TPO-R | |
Mouse | |
Protein G | |
RUO | |
4352 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |