missing translation for 'onlineSavingsMsg'
Learn More

MRE11 Monoclonal Antibody (12D7), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA123253

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11521480

  • 469.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: HeLa, Jurkat, Raji, NCI-H929, 293T, human Mre11-transfected 293T, PC-12.

MRE11 Monoclonal antibody specifically detects MRE11 in Human, Mouse, Rat samples. It is validated for ELISA, Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot.
Spezifikation

Spezifikation

MRE11
Monoclonal
0.45 mg/mL
PBS with no preservative
P49959, Q61216, Q9JIM0
Mre11, Mre11a
Amino acids 182-582 of Mre11 expressed in E. coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
ELISA, Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot
12D7
Unconjugated
Mre11
ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C
Mouse
Protein G
RUO
17535, 4361, 64046
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote