missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRE11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141583
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: 293 whole cell lysate.
MRE11 Polyclonal antibody specifically detects MRE11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MRE11 | |
Polyclonal | |
Unconjugated | |
Mre11 | |
ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C | |
Rabbit | |
Protein G | |
RUO | |
4361 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P49959 | |
Mre11 | |
Synthetic peptide corresponding to a portion of human MRE11. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |