missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRFAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545688
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: KTQVEASEES ALNHLQNPGD AAEGRAAKRC EKAEEKAKEI AKMAEMLVEL Sequence homology: Cow: 100%; Dog: 86%; Human: 100%; Pig: 100%
MRFAP1 Polyclonal antibody specifically detects MRFAP1 in Human samples. It is validated for Western Blot.Spezifikation
MRFAP1 | |
Polyclonal | |
Unconjugated | |
MRFAP1 | |
9130413I22Rik; AL022827; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; Morf4 family associated protein 1 like 1; Morf4 family associated protein 1-like 1; MORF4 family-associated protein 1; MORF4 family-associated protein 1-like 1; MRFAP1; MRFAP1L1; PAM14; PGR1; PP784; protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; Protein PGR1; T-cell activation protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
93621 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y605 | |
MRFAP1 | |
Synthetic peptide directed towards the C-terminal of human MRFAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |