missing translation for 'onlineSavingsMsg'
Learn More

MRGD Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA533942

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15302701

  • 514.00 € / 50 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon, Monkey (94%).

MRGD Polyclonal antibody specifically detects MRGD in Human samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

MRGD
Polyclonal
Unconjugated
MRGPRD
Beta-alanine receptor; G protein-coupled receptor MrgD; Gm499; G-protein coupled receptor TGR7; MAS related GPR family member D; mas-related G protein-coupled MRGD; Mas-related G protein-coupled receptor d; MAS-related gene D; MAS-related GPR, member D; mas-related G-protein coupled receptor member D; MAS-related G-protein coupled receptor, member D; Mgrd; MrgD; MRGD G protein-coupled receptor; MRGPRD; TGR7
Rabbit
Antigen affinity chromatography
RUO
116512
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q8TDS7
MRGPRD
Synthetic 18 amino acid peptide from 2nd extracellular domain of human MRGPRD.
50 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote