missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533945
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%).
MRGPRE Polyclonal antibody specifically detects MRGPRE in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
MRGPRE | |
Polyclonal | |
Unconjugated | |
MRGPRE | |
C130069N09Rik; Ebrt3; Evolutionary breakpoint transcript 3 protein; G protein-coupled receptor 167; G protein-coupled receptor MrgE; GPR167; G-protein coupled receptor 167; MAS related GPR family member E; mas-related G protein-coupled MRGE; Mas-related G protein-coupled receptor F; MAS-related gene E; MAS-related GPR, member E; mas-related G-protein coupled receptor member E; MAS-related G-protein coupled receptor, member E; MGC138408; Mgrf; MrgE; MRGPRE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q86SM8 | |
MRGPRE | |
Synthetic 19 amino acid peptide from C-terminus of human MRGPRE. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |