Learn More
MRGPRF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532928
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset, Dog, Bovine, Horse (94%); Mouse, Rat, Hamster, Elephant, Panda, Pig (88%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
MRGPRF Polyclonal antibody specifically detects MRGPRF in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
MRGPRF | |
Polyclonal | |
Unconjugated | |
MRGPRF | |
BC019711; G protein coupled receptor (RTA); G protein coupled receptor Rca; G protein-coupled receptor 140; G protein-coupled receptor 168; G protein-coupled receptor MrgF; G protein-coupled receptor RTA; G-p; GPR140; GPR168; G-protein coupled receptor 140; G-protein coupled receptor 168; G-protein coupled receptor RTA; MAS related GPR family member F; mas-related G protein-coupled MRGF; Mas-related G protein-coupled receptor c; mas-related gene; mas-related gene F protein; MAS-related GPR, member F; mas-related G-protein coupled receptor member F; MAS-related G-protein coupled receptor, member F; MGC21621; Mgrc; MRGF; MRGPRF; PSEC0142; Rta; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96AM1 | |
MRGPRF | |
Synthetic 18 amino acid peptide from N-Terminus of human MRGPRF. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |