missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533949
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Monkey (100%) Gibbon (93%).
MRGPRX1 Polyclonal antibody specifically detects MRGPRX1 in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
MRGPRX1 | |
Polyclonal | |
Unconjugated | |
Mrgprx1 | |
G protein-coupled receptor MRGX1; G protein-coupled receptor SNSR; G protein-coupled receptor SNSR3; GPCR; MAS related GPR family member X1; Mas-related G protein-coupled receptor g2; MAS-related GPR, member C11; MAS-related GPR, member X1; mas-related G-protein coupled receptor member C11; Mas-related G-protein coupled receptor member X1; MGRG2; Mrgc; Mrgc11; Mrgprc; Mrgprc11; mrgprx1; MRGX1; sensory neuron-specific G protein-coupled receptor 1; sensory neuron-specific G-protein coupled receptor 1; Sensory neuron-specific G-protein coupled receptor 3/4; Snsr; Snsr1; SNSR3; SNSR4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
259249 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96LB2 | |
Mrgprx1 | |
Synthetic 14 amino acid peptide from N-terminal extracellular domain of human MRGPRX1 / MRGX1. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |