missing translation for 'onlineSavingsMsg'
Learn More

MRPL1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA530806

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13415317

  • 451.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA5-30806 targets MRPL1 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-30806 immunogen is recombit fragment corresponding to a region within amino acids 128 and 325 of Human MRPL1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L1 ribosomal protein family.
Spezifikation

Spezifikation

MRPL1
Polyclonal
Unconjugated
Q9BYD6, Q99N96
MRPL1
Recombit fragment corresponding to a region within amino acids 128 and 325 of Human MRPL1
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
0.84 mg/mL
MRPL1
BM022, L1MT, MRP-L1, 39S ribosomal protein L1, mitochondrial
Rabbit
Antigen affinity chromatography
RUO
65008, 94061
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote