Learn More
MRPL10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531244
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, Raji. Predicted reactivity: Human (99%), Rat (84%), Rhesus Monkey (96%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MRPL10 Polyclonal antibody specifically detects MRPL10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MRPL10 | |
Polyclonal | |
Unconjugated | |
MRPL10 | |
0610040E02Rik; 39S ribosomal protein L10, mitochondrial; 39S ribosomal protein L8, mitochondrial; C78440; L10MT; L8mt; Mitochondrial large ribosomal subunit protein uL10m; mitochondrial ribosomal protein L10; MRPL10; MRP-L10; MRPL8; MRP-L8; Rpml8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124995 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q7Z7H8 | |
MRPL10 | |
Recombinant protein fragment corresponding to a region within amino acids 58 and 230 of Human MRPL10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |