Learn More
MRPL12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530159
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (84%), Rat (82%), Rhesus Monkey (94%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MRPL12 Polyclonal antibody specifically detects MRPL12 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MRPL12 | |
Polyclonal | |
Unconjugated | |
MRPL12 | |
0610034O11Rik; 1500031N16Rik; 39S ribosomal protein L12, mitochondrial; 5c5-2; FLJ60124; hypothetical protein LOC550391; im:6893415; L12mt; MGC8610; Mitochondrial large ribosomal subunit protein bL12m; mitochondrial ribosomal protein L12; mitochondrial ribosomal protein L7/12; MRPL12; MRP-L12; MRP-L31/34; MRPL7; MRPL7/L12; ribosomal protein, mitochondrial, L12; Rpm12; RPML12; zgc:112445 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56282, 6182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.56 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P52815, Q9DB15 | |
MRPL12 | |
Recombinant fragment corresponding to a region within amino acids 1 and 198 of Human MRPL12 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |