missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538968
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53 kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified. [provided by RefSeq, Jul 2008].Spezifikation
MRPL47 | |
Polyclonal | |
Unconjugated | |
MRPL47 | |
NCM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57129, 74600 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HD33, Q8K2Y7 | |
MRPL47 | |
A synthetic peptide derived from the internal region of human MRPL47 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |