Learn More
MRPL48 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544624
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: KKGKVEVRAI NLGTDYEYGV LNIHLTAYDM TLAESYAQYV HNLCNSLSIK Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
MRPL48 Polyclonal antibody specifically detects MRPL48 in Human samples. It is validated for Western Blot.Spezifikation
MRPL48 | |
Polyclonal | |
Unconjugated | |
MRPL48 | |
1810030E20Rik; 2610028L11Rik; 39S ribosomal protein L48, mitochondrial; CGI-118; D4Ertd786e; HSPC290; L48mt; MGC13323; mitochondrial 39S ribosomal protein L48-like protein; Mitochondrial large ribosomal subunit protein mL48; mitochondrial ribosomal protein L48; Mrpl48; MRP-L48; Unknown (protein for MGC:133957) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51642 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96GC5 | |
MRPL48 | |
Synthetic peptide directed towards the middle region of human MRPL48 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |