missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS18C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538951
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 8p, 12p, 15q, and 22q. [provided by RefSeq, Jul 2008].Spezifikation
MRPS18C | |
Polyclonal | |
Unconjugated | |
MRPS18C | |
MRP-S18-1, MRPS18-1, 28S ribosomal protein S18-1, mitochondrial, 28S ribosomal protein S18c, mitochondrial, MRP-S18-c, S18mt-c, mitochondrial ribosomal protein S18-1, mrps18-c | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51023, 68735 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y3D5, Q8R2L5 | |
MRPS18C | |
A synthetic peptide derived from the internal region of human MRPS18C | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |