missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRVI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532111
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: MCF-7. Predicted reactivity: Rat (86%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MRVI1 Polyclonal antibody specifically detects MRVI1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MRVI1 | |
Polyclonal | |
Unconjugated | |
MRVI1 | |
BB115629; inositol 1,4,5-triphosphate receptor associated 1; inositol 1,4,5-triphosphate receptor associated 1; protein MRVI1; inositol 1,4,5-triphosphate receptor-associated cGMP kinase substrate; inositol 1,4,5-trisphosphate receptor-associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate A; IP3 receptor associated cGMP kinase substrate B; IP3R-associated cGMP kinase substrate; IRAG; IRAG1; JAW1L; JAW1-related protein MRVI1; MRV integration site 1; MRVI1; murine retrovirus integration site 1 homolog; murine retrovirus integration site 1 protein; Protein MRVI1; retroviral integration site 1; Ris1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10335 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y6F6 | |
MRVI1 | |
Recombinant protein fragment corresponding to a region within amino acids 404 and 547 of Human MRVI1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |