missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Thermo Scientific PA19024
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD20 Polyclonal antibody specifically detects CD20 in Human samples. It is validated for Western Blot.Spezifikation
CD20 | |
Polyclonal | |
Unconjugated | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
931 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P11836 | |
Ms4a1 | |
Synthetic peptide corresponding to the C terminus of MS4A1, amino acids QDQESSPIENDSSP. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |