Learn More
MS4A15 Monoclonal Antibody (1D4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523183
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: MVRRGHVGIF FIEGGVPFWG GACFIISGSL SVAAEKNHTS CLVRSSLGTN ILSVMAAFAG TAILLMDFGV TNRDVDRGYL AVLTIFTVLE FFTAVIAMHF GCQAIHAQAS APVIFLPNAF SADFNIPSPA ASAPPAYDNV AYAQGVV
MS4A15 Monoclonal antibody specifically detects MS4A15 in Human samples. It is validated for ELISA.Spezifikation
MS4A15 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8N5U1 | |
MS4A15 | |
MS4A15 (AAH31610.1, 1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
1D4 | |
Unconjugated | |
MS4A15 | |
membrane spanning 4-domains A15; membrane-spanning 4-domains subfamily A member 15; membrane-spanning 4-domains, subfamily A, member 15; MS4A15 | |
Mouse | |
Affinity chromatography | |
RUO | |
219995 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |