missing translation for 'onlineSavingsMsg'
Learn More

MSH3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532508

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13417027

  • 474.00 € / 500 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is rectal carcinoma.

MSH3 Polyclonal antibody specifically detects MSH3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

MSH3
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P20585
MSH3
Synthetic peptide derived from internal region of human MSH3 protein.
500 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
MSH3
D13Em1; Divergent upstream protein; DNA mismatch repair protein Msh3; DUC1; DUG; DUP; hMSH3; Mismatch repair protein 1; MRP1; MSH3; mutS homolog 3; mutS homolog 3 (E. coli); protein repair-1; Protein repair-3; REP-1; Rep3; Rep-3
Rabbit
Antigen affinity chromatography
RUO
4437
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote