Learn More
MSH5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541990
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DENMTRFLGK LASQEHREPK RPEIIFLPSV DFGLEISKQR LLSGNYSFIP Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 100%
MSH5 Polyclonal antibody specifically detects MSH5 in Human samples. It is validated for Western Blot.Spezifikation
MSH5 | |
Polyclonal | |
Unconjugated | |
MSH5 | |
DKFZp434C1615; G7; hMSH5; MGC2939; MSH5; Mut5; mutS homolog 5; mutS homolog 5 (E. coli); mutS protein homolog 5; MUTSH5; NG23 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4439 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43196 | |
MSH5 | |
Synthetic peptide directed towards the N-terminal of human MSH5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |