Learn More
MSH6 Monoclonal Antibody (3A10H7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515392
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15392 targets MSH6 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15392 immunogen is purified recombinant fragment of MSH6 expressed in E. Coli. <br>.
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
MSH6 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P52701 | |
MSH6 | |
Purified recombinant fragment of MSH6 expressed in E. Coli. | |
RUO | |
2956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
3A10H7 | |
Unconjugated | |
MSH6 | |
AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |