Learn More
MSH6 Monoclonal Antibody (SP93), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA516380
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is rectal carcinoma.
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MSH6 | |
Monoclonal | |
Unconjugated | |
MSH6 | |
AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
Rabbit | |
Protein A/G | |
RUO | |
2956 | |
4° C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP93 | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P52701 | |
MSH6 | |
Synthetic peptide corresponding to internal region of human MSH6 protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |