Learn More
MSH6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529348
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, Neuro2A, GL261. Predicted reactivity: Mouse (88%), Rhesus Monkey (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MSH6 Polyclonal antibody specifically detects MSH6 in Human, Mouse, Zebrafish samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
MSH6 | |
Polyclonal | |
Unconjugated | |
MSH6 | |
AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17688, 260437, 2956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P52701, P54276 | |
MSH6 | |
Recombinant fragment corresponding to a region within amino acids 825 and 1110 of Human MSH6 | |
100 μL | |
Primary | |
Human, Mouse, Zebrafish | |
Antibody | |
IgG |