Learn More
Moesin Monoclonal Antibody (2C12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517130
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17130 targets MSN in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-17130 immunogen is purified recombinant fragment of human MSN expressed in E. Coli. MA5-17130 detects MSN which has a predicted molecular weight of approximately 67.8kDa.
Moesin Monoclonal antibody specifically detects Moesin in Human, Non-human Primate samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
Moesin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P26038 | |
MSN | |
Purified recombinant fragment of human MSN expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
2C12 | |
Unconjugated | |
MSN | |
C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN | |
Mouse | |
Protein G | |
RUO | |
4478 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |