missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRB2 Monoclonal Antibody (3F12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA521370
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: KEAGMYHCVC CDSPLFSSEK KYCSGTGWPS FSEAHGTSGS DESHTGILRR LDTSLGSART EVVCKQCEAH LGHVFPDGPG PNGQRFCINS VALKFKPRKH
MSRB2 Monoclonal antibody specifically detects MSRB2 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
MSRB2 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9Y3D2 | |
MSRB2 | |
MSRB2 (NP_036360, 102 a.a. ∽ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
3F12 | |
Unconjugated | |
MSRB2 | |
2310050L06Rik; CBS1; CBS-1; CGI-131; methionine sulfoxide reductase B2; methionine-R-sulfoxide reductase B2, mitochondrial; Mrsb; Msrb; MsrB2; PILB; pilin-like transcription factor | |
Mouse | |
Affinity chromatography | |
RUO | |
22921 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |