missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ATP6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537872
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
MT-ATP6 Polyclonal antibody specifically detects MT-ATP6 in Human samples. It is validated for Western Blot.Spezifikation
MT-ATP6 | |
Polyclonal | |
Unconjugated | |
MT-ATP6 | |
ATP synthase F0 subunit 6; ATP synthase subunit a; Atp6; ATPase6; F-ATPase protein 6; mitochondrially encoded ATP synthase 6; Mtatp6; MT-ATP6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4508 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P00846 | |
MT-ATP6 | |
Peptide with sequence C-PTSKYLINNRLITTQ, from the internal region of ATP6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |