missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ATP6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537129
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 27 kDa. Purity is >95% by SDS-PAGE.
MT-ATP6 Polyclonal antibody specifically detects MT-ATP6 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
MT-ATP6 | |
Polyclonal | |
Unconjugated | |
MT-ATP6 | |
ATP synthase F0 subunit 6; ATP synthase subunit a; Atp6; ATPase6; F-ATPase protein 6; mitochondrially encoded ATP synthase 6; Mtatp6; MT-ATP6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17705, 26197, 4508 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P00846, P00848, P05504 | |
MT-ATP6 | |
A synthetic peptide corresponding to residues in human MT-ATP6 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |