Learn More
MTCH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542964
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NNCGLQAGLP PYSPVFKSWI HCWKYLSVQG QLFRGSSLLF RRVSSGSCFA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
MTCH1 Polyclonal antibody specifically detects MTCH1 in Human samples. It is validated for Western Blot.Spezifikation
MTCH1 | |
Polyclonal | |
Unconjugated | |
MTCH1 | |
2310034O17Rik; AI255158; AU018396; C77849; cell proliferation-inducing protein 60; CGI-64; mitochondrial carrier 1; mitochondrial carrier homolog 1; mitochondrial carrier homolog 1 (C. elegans); mitochondrial carrier-like protein 1; Mtch1; PIG60; presenilin-associated protein; PSAP; SLC25A49; solute carrier family 25, member 49; UNQ1871/PRO4314 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23787 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZJ7 | |
MTCH1 | |
Synthetic peptide directed towards the C-terminal of human MTCH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |