missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525406
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MTCH2 Polyclonal antibody specifically detects MTCH2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MTCH2 | |
Polyclonal | |
Unconjugated | |
Mtch2 | |
2310034D24Rik; 4930539J07Rik; HSPC032; Met-induced mitochondrial protein; MIMP; mitochondrial carrier 2; mitochondrial carrier homolog 2; mitochondrial carrier homolog 2 (C. elegans); Mtch2; SLC25A50; solute carrier family 25, member 50 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23788, 56428 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q791V5, Q9Y6C9 | |
Mtch2 | |
KLH conjugated synthetic peptide between 75-103 amino acids from the N-terminal region of human MTCH2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |