missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534530
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is NIH-3T3 cell lysate. PA5-34530 can be used with blocking peptide PEP-1573.
MTERFD1 Polyclonal antibody specifically detects MTERFD1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
MTERFD1 | |
Polyclonal | |
Unconjugated | |
MTERF3 | |
2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51001, 66410 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8R3J4, Q96E29 | |
MTERF3 | |
A 17 amino acid peptide near the carboxy terminus of human MTERFD1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |