missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535932
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (98%), Rat (96%), Chicken (90%).
MTF2 Polyclonal antibody specifically detects MTF2 in Human samples. It is validated for Immunocytochemistry.Spezifikation
MTF2 | |
Polyclonal | |
Unconjugated | |
Mtf2 | |
9230112N11Rik; AA537621; C76717; dJ976O13.2; hPCl2; M96; Metal regulatory transcription factor 2; metal response element binding transcription factor 2; metal response element DNA-binding protein M96; metal response element-binding transcription factor 2; metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; mPCl2; MTF2; PCL2; polycomb-like 2; polycomb-like protein 2; putative DNA binding protein; RGD1304727; RP5-976O13.1; TDRD19A; tudor domain containing 19A; zinc-regulated factor 1; ZiRF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y483 | |
Mtf2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 199 of MTF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |