Learn More
MTHFD2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544491
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: TGIQTFGKNV VVAGRSKNVG MPIAMLLHTD GEHERPGGDA TVTIAHRYTP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
MTHFD2L Polyclonal antibody specifically detects MTHFD2L in Human samples. It is validated for Western Blot.Spezifikation
MTHFD2L | |
Polyclonal | |
Unconjugated | |
MTHFD2L | |
1110019K23Rik; C630010D07Rik; FLJ13105; LOW QUALITY PROTEIN: probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; methylenetetrahydrofolate dehydrogenase 2-like; MGC45532; MGC72244; MTHFD2L; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; RGD1310879; tetrahydrofolate dehydrogenase/cyclohydrolase; Unknown | |
Rabbit | |
Affinity Chromatography | |
RUO | |
441024 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H903 | |
MTHFD2L | |
Synthetic peptide directed towards the middle region of human MTHFD2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |