Learn More
MTHFS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530565
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HepG2. Predicted reactivity: Mouse (85%), Rat (85%), Rhesus Monkey (95%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MTHFS Polyclonal antibody specifically detects MTHFS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MTHFS | |
Polyclonal | |
Unconjugated | |
Mthfs | |
1110034I12Rik; 2310020H23Rik; 5, 10-methenyltetrahydrofolate synthetase; 5,10-methenyl-tetrahydrofolate synthetase; 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase); 5-formyltetrahydrofolate cyclo-ligase; AI119695; FLJ30410; HsT19268; methenyl-THF synthetase; Mthfs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.75 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P49914 | |
Mthfs | |
Recombinant fragment corresponding to a region within amino acids 7 and 203 of Human MTHFS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |