missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTM1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA517972
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and mouse based on sequence homology.
MTM1 Polyclonal antibody specifically detects MTM1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MTM1 | |
Polyclonal | |
Unconjugated | |
MTM1 | |
AF073996; CG2; CNM; fb19c01; hypothetical protein LOC560881; im:6903724; mKIAA4176; Mtm; MTM1; MTMX; myotubularin; myotubularin 1; Myotubularin-like protein; phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; wu:fb19c01; X-linked myotubular myopathy gene 1; XLMTM; zgc:123266 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
17772, 4534 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13496, Q9Z2C5 | |
MTM1 | |
Synthetic peptide sequence (SSPSQMMPHVQTHF) corresponding to the C-terminus amino acids of MTM1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |