missing translation for 'onlineSavingsMsg'
Learn More

MTMR12 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531375

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13405897

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, Raji, mouse brain. Predicted reactivity: Mouse (90%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MTMR12 Polyclonal antibody specifically detects MTMR12 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

MTMR12
Polyclonal
Unconjugated
MTMR12
3Pap; 3-PAP; 3-phosphatase adapter protein; 3-phosphatase adapter subunit; 4932703C11; C730015A02Rik; im:7160376; Inactive phosphatidylinositol 3-phosphatase 12; Kiaa1682; mKIAA1682; Mtmr12; myotubularin related protein 12; myotubularin-related protein 12; Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol 3-phosphatase-associated protein; phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit; phosphatidylinositol-3-phosphatase associated protein; phosphatidylinositol-3-phosphatase-associated protein; phosphatidylinositol-3-phosphate associated protein; Pip3ap; si:dkey-207e22.3
Rabbit
Antigen affinity chromatography
RUO
268783, 54545
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q80TA6, Q9C0I1
MTMR12
Recombinant protein fragment corresponding to a region within amino acids 453 and 656 of Human MTMR12
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote