Learn More
MTMR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527748
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Mouse (98%), Rat (96%), Zebrafish (82%), Chicken (95%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MTMR2 Polyclonal antibody specifically detects MTMR2 in Human samples. It is validated for Western Blot.Spezifikation
MTMR2 | |
Polyclonal | |
Unconjugated | |
MTMR2 | |
6030445P13Rik; CMT4B; CMT4B1; KIAA1073; Mtmr2; myotubularin related protein 2; myotubularin-related protein 2; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; phosphoinositide-3-phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13614 | |
MTMR2 | |
Recombinant fragment corresponding to a region within amino acids 417 and 643 of Human MTMR2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |