missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTNR1A Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519109
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and mouse based on sequence homology.
MTNR1A Polyclonal antibody specifically detects MTNR1A in Rat samples. It is validated for Western Blot.Spezifikation
MTNR1A | |
Polyclonal | |
Unconjugated | |
MTNR1A | |
mel1a melatonin receptor; mel1a receptor; Mel-1A-R; melatonin receptor 1A; melatonin receptor type 1A; MelR; MR; MT1; Mtnr1a | |
Goat | |
Antigen affinity chromatography | |
RUO | |
114211 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P49218 | |
MTNR1A | |
Synthetic peptide sequence (SLKYDKLYSSKNS) corresponding to the internal amino acids of MTNR1A | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |