Learn More
mTOR Recombinant Rabbit Monoclonal Antibody (7H13L18), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701483
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
mTOR Recombinant Monoclonal antibody specifically detects mTOR in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
mTOR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
Recombinant protein corresponding to amino acids 1126–1411 of human mTOR | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Western Blot | |
7H13L18 | |
Unconjugated | |
P42345 | |
Rabbit | |
Protein A | |
RUO | |
2475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |