Learn More
CRYM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543759
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: AHINAVGASR PDWRELDDEL MKEAVLYVDS QEAALKESGD VLLSGAEIFA Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%
CRYM Polyclonal antibody specifically detects CRYM in Human samples. It is validated for Western Blot.Spezifikation
CRYM | |
Polyclonal | |
Unconjugated | |
CRYM | |
CDK108; Crym; crystallin mu; crystallin, mu; DFNA40; ketimine reductase mu-crystallin; mu-crystallin homolog; Mu-crystallin-like protein; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; similar to Mu-crystallin homolog (NADP-regulated thyroid-hormone binding protein); THBP; thiomorpholine-carboxylate dehydrogenase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1428 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14894 | |
CRYM | |
Synthetic peptide directed towards the middle region of human CRYM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |