missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523249
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MUC13 Polyclonal antibody specifically detects MUC13 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MUC13 | |
Polyclonal | |
Unconjugated | |
MUC13 | |
114/A10; 14/A10; AI159736; Cell surface antigen 114/A10; down-regulated in colon cancer 1; DRCC1; Lrrp; Ly64; Lymphocyte antigen 64; Muc-123; MUC13; MUC-13; mucin 13; mucin 13, cell surface associated; mucin 13, epithelial transmembrane; mucin-13; NJ-1; RECC; UNQ6194/PRO20221 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9H3R2 | |
MUC13 | |
A portion of amino acids 480-512 of human Mucin 13 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |