missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Monoclonal Antibody (996/1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512345
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-12345 targets Mucin 2 in IHC (P), ICC/IF and FACS applications and shows reactivity with human and mouse samples. The MA5-12345 immunogen is mUC2 tandem repeat peptide.
MUC2 Monoclonal antibody specifically detects MUC2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).Spezifikation
MUC2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q02817, Q80Z19 | |
MUC2 | |
MUC2 tandem repeat peptide | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
996/1 | |
Unconjugated | |
MUC2 | |
2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
Mouse | |
Protein G | |
RUO | |
17831, 4583 | |
4° C | |
Liquid |