missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC3 Monoclonal Antibody (1143/B7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512350
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-12350 targets Mucin 3 in IHC (P) applications and shows reactivity with Human samples. The MA5-12350 immunogen is mUC3 tandem repeat peptide.
MUC3 Monoclonal antibody specifically detects MUC3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
MUC3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q02505 | |
MUC3A | |
MUC3 tandem repeat peptide | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin) | |
1143/B7 | |
Unconjugated | |
MUC3A | |
A630081J09Rik; intestinal mucin; Intestinal mucin-3A; MUC3; MUC3A; MUC-3A; mucin 3; mucin 3, intestinal; mucin 3A; mucin 3A, cell surface associated; Mucin-3; mucin-3A; RIKEN cDNA A630081J09 gene | |
Mouse | |
Protein A | |
RUO | |
4584 | |
4° C | |
Liquid |