missing translation for 'onlineSavingsMsg'
Learn More

MUC5AC Monoclonal Antibody (45M1), Biotin, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA512175

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11304593

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

MA5-12175 targets Mucin 5AC in IHC (P) applications and shows reactivity with Chicken, Feline, Human, mouse, Non-human primate, Porcine, Rabbit, Hedgehog, and Rat samples. The MA5-12175 immunogen is m1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient.

MUC5AC Monoclonal antibody specifically detects MUC5AC in Chicken, Feline, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat, Rodent samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

MUC5AC
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P98088
MUC5AC, MUC5ACL
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
500 μL
Primary
Chicken, Feline, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat, Rodent
Antibody
IgG1 κ
Immunohistochemistry (Paraffin)
45M1
Biotin
MUC5AC
2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Mouse
Protein G
RUO
100170143, 100859916, 105261014, 110259210, 17833, 4586, 682837
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote