missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534612
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with gorilla based on sequence homology.
MUC5AC Polyclonal antibody specifically detects MUC5AC in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
MUC5AC | |
Polyclonal | |
Unconjugated | |
MUC5AC | |
2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4586 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 10mg/mL BSA and 0.02% sodium azide; pH 7.4 | |
P98088 | |
MUC5AC | |
Synthetic peptide conjugated to KLH derived from within residues 5000 to the C-terminus of Human Mucin 5AC. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |