missing translation for 'onlineSavingsMsg'
Learn More

MUM1 Monoclonal Antibody (4G10), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA515639

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11375603

  • 463.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-15639 targets MUM1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15639 immunogen is purified recombinant fragment of human MUM1 expressed in E. Coli.<br>. MA5-15639 detects MUM1 which has a predicted molecular weight of approximately 78.6kDa.

MUM1 Monoclonal antibody specifically detects MUM1 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

MUM1
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
Q2TAK8
MUM1
Purified recombinant fragment of human MUM1 expressed in E. Coli.
RUO
84939
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry, Western Blot
4G10
Unconjugated
MUM1
2610019J09; 9430059D04Rik; EXPAND1; FLJ14868; FLJ22283; HSPC211; melanoma associated antigen (mutated) 1; melanoma ubiquitous mutated protein; MGC131891; MGC163315; Mum1; MUM-1; Mutated melanoma-associated antigen 1; protein expandere; PWWP domain-containing DNA repair factor 3A; PWWP domain-containing protein MUM1; Pwwp3a; UBE-1C2
Mouse
100 μL
Primary
Human
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote